Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006339.2 | NP_006330.2 | 5 | 448 | ||
XM_017026144.1 | XP_016881633.1 | 5 | 455 | |||
GenBank mRNA | AF072165.1 | - | 2 | 354 | ||
AF072836.1 | - | 2 | 92 | |||
AF146223.1 | - | 4 | 390 | |||
AF288679.1 | - | 4 | 380 | |||
AF318366.1 | - | 3 | 1271 | |||
AF331191.1 | - | 4 | 380 | |||
AK090733.1 | - | 5 | 342 | |||
AK289500.1 | - | 5 | 436 | |||
AK314201.1 | - | 5 | 422 | |||
AL355691.1 | - | 3 | 346 | |||
AL355703.1 | - | 4 | 392 | |||
AL355709.1 | - | 4 | 390 | |||
BC002552.2 | - | 5 | 438 | 3138919 | ||
BC003505.1 | - | 5 | 436 | 3609369 | ||
BC004408.1 | - | 4 | 386 | 3638942 | ||
BC021585.1 | - | 3 | 350 | 4873122 | ||
CR456754.1 | - | 2 | 74 |