Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175039.3 | NP_778204.1 | 3 | 454 | ||
NM_175040.3 | NP_778205.1 | 2 | 367 | |||
XM_011518527.2 | XP_011516829.1 | 4 | 546 | |||
XM_011518529.2 | XP_011516831.1 | 4 | 549 | |||
GenBank mRNA | AB035172.1 | - | 2 | 199 | ||
AJ271734.1 | - | 3 | 469 | |||
AK000600.1 | - | 3 | 450 | |||
AK292049.1 | - | 2 | 358 | |||
BC036705.2 | - | 3 | 360 | 4309208 | ||
CB126299.1 | - | 2 | 367 |