Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016466.5 | NP_057550.3 | 2 | 179 | ||
GenBank mRNA | AF151034.1 | - | 2 | 159 | ||
AK312193.1 | - | 2 | 176 | |||
BC031303.2 | - | 2 | 171 | 5284947 | ||
DA769410.1 | - | 2 | 176 |