Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040437.2 | NP_001035527.1 | 1 | 172 | ||
NM_001040438.2 | NP_001035528.1 | 1 | 172 | |||
NM_001287482.1 | NP_001274411.1 | 1 | 172 | |||
NM_001287483.1 | NP_001274412.1 | 1 | 172 | |||
NM_001287484.1 | NP_001274413.1 | 1 | 172 | |||
GenBank mRNA | AJ249731.1 | - | 1 | 181 | ||
AJ249732.1 | - | 1 | 181 | |||
AK311989.1 | - | 1 | 171 | |||
AL521625.3 | - | 1 | 158 | |||
AL556045.3 | - | 1 | 138 | |||
BC000706.1 | - | 1 | 153 | 3349773 | ||
BC017660.1 | - | 2 | 70 | 4049404 | ||
BC066330.1 | - | 1 | 171 | |||
BG109213.1 | - | 1 | 100 | |||
BI114651.1 | - | 1 | 149 | |||
BJ996767.1 | - | 1 | 172 | |||
BQ920527.1 | - | 1 | 156 | |||
CD249104.1 | - | 1 | 187 | |||
W52524.1 | - | 1 | 159 |