Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320546.1 | NP_001307475.1 | 2 | 754 | ||
NM_001320547.1 | NP_001307476.1 | 2 | 125 | |||
NM_018031.4 | NP_060501.3 | 2 | 540 | |||
GenBank mRNA | AF099100.1 | - | 2 | 209 | ||
AK001080.1 | - | 2 | 209 | |||
AK092079.1 | - | 2 | 208 | |||
AK225653.1 | - | 2 | 203 | |||
AK295145.1 | - | 2 | 540 | |||
AK304117.1 | - | 2 | 125 | |||
AK309913.1 | - | 2 | 420 | |||
BC002826.2 | - | 2 | 195 | |||
BC101707.1 | - | 2 | 199 | 8069213 | ||
BC113467.1 | - | 2 | 199 | 8322519 | ||
BC143486.1 | - | 2 | 199 | |||
DC300417.1 | - | 2 | 125 | |||
DC377068.1 | - | 2 | 444 | |||
DC389885.1 | - | 2 | 293 | |||
DC400613.1 | - | 2 | 398 | |||
DC408104.1 | - | 2 | 321 |