Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318395.1 | NP_001305324.1 | 3 | 636 | ||
NM_031948.4 | NP_114154.1 | 4 | 799 | |||
XM_011522689.2 | XP_011520991.1 | 2 | 528 | |||
GenBank mRNA | AB056161.1 | - | 4 | 458 | ||
AJ306593.1 | - | 4 | 427 | |||
AK055576.1 | - | 1 | 1400 | |||
AK310214.1 | - | 3 | 311 | |||
AY030095.1 | - | 4 | 408 | |||
AY359106.1 | - | 4 | 443 | |||
BC034294.2 | - | 4 | 463 | 4752547 | ||
BC036912.2 | - | 3 | 647 |