Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032880.4 | NP_116269.3 | 2 | 473 | ||
XM_011542319.2 | XP_011540621.1 | 2 | 430 | |||
XM_017002604.1 | XP_016858093.1 | 2 | 119 | |||
GenBank mRNA | AK075316.1 | - | 2 | 473 | ||
AK294121.1 | - | 2 | 42 | |||
BC007618.1 | - | 2 | 393 | 3355289 |