Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052844.3 | NP_443076.2 | 2, 3 | 558 | ||
XM_011519179.2 | XP_011517481.1 | 2, 3 | 492 | |||
GenBank mRNA | AL555205.3 | - | 2, 3 | 365 | ||
BC001614.1 | - | 2, 3 | 393 | |||
BC011874.2 | - | 2, 3 | 495 | 3161070 | ||
BG177634.1 | - | 2, 3 | 402 | |||
BI910783.1 | - | 3, 4 | 653 |