Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256171.1 | NP_001243100.1 | 3 | 339 | ||
NM_001256172.1 | NP_001243101.1 | 3 | 339 | |||
NM_001256173.1 | NP_001243102.1 | 2 | 212 | |||
NM_003429.4 | NP_003420.2 | 3 | 339 | |||
NR_045830.1 | 3 | 352 | ||||
XM_011528263.2 | XP_011526565.1 | 3 | 365 | |||
GenBank mRNA | BC008688.1 | - | 3 | 295 | ||
BC068066.1 | - | 3 | 316 | |||
BM755583.1 | - | 3 | 338 | |||
BP199573.1 | - | 3 | 339 | |||
DB119881.1 | - | 2 | 161 | |||
DC358791.1 | - | 3 | 307 | |||
DN994243.1 | - | 3 | 326 | |||
M61866.1 | - | 3 | 303 | |||
U35376.1 | - | 3 | 309 |