Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175881.3 | NP_787077.1 | 2 | 426 | ||
XM_006720414.3 | XP_006720477.1 | 2 | 1508 | |||
GenBank mRNA | BC039862.1 | - | 2 | 426 | 5744333 |