Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG2 SIRNA Assays ›
Gene Symbol:
DLG2
Gene Name:
discs large MAGUK scaffold protein 2
Gene Aliases:
PPP1R58, PSD-93, PSD93, chapsyn-110
Chromosome Location:
Chr.11: 83455009 - 85628534 on Build GRCh38
UniGene ID:
Hs.367656
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
46571
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142699.1
NP_001136171.1
28
6075
NM_001142700.1
NP_001136172.1
20
5110
NM_001142702.1
NP_001136174.1
11
4152
NM_001206769.1
NP_001193698.1
22
5759
NM_001300983.1
NP_001287912.1
23
5697
NM_001364.3
NP_001355.2
23
5751
XM_005273810.4
XP_005273867.1
22
5394
XM_005273811.4
XP_005273868.1
22
5340
XM_011544778.2
XP_011543080.1
29
7366
XM_011544780.2
XP_011543082.1
29
7312
XM_011544782.2
XP_011543084.1
24
5837
XM_011544788.2
XP_011543090.1
21
5184
XM_017017254.1
XP_016872743.1
30
7408
XM_017017255.1
XP_016872744.1
29
7369
XM_017017256.1
XP_016872745.1
29
6898
XM_017017257.1
XP_016872746.1
28
7830
XM_017017258.1
XP_016872747.1
27
7006
XM_017017259.1
XP_016872748.1
27
6727
XM_017017260.1
XP_016872749.1
26
7659
XM_017017261.1
XP_016872750.1
28
7156
XM_017017262.1
XP_016872751.1
23
5861
XM_017017263.1
XP_016872752.1
25
5879
XM_017017264.1
XP_016872753.1
24
6056
XM_017017265.1
XP_016872754.1
24
6053
XM_017017266.1
XP_016872755.1
22
5763
XM_017017267.1
XP_016872756.1
24
5961
XM_017017268.1
XP_016872757.1
24
5782
XM_017017269.1
XP_016872758.1
24
6483
XM_017017270.1
XP_016872759.1
24
5783
XM_017017271.1
XP_016872760.1
23
5960
XM_017017272.1
XP_016872761.1
23
5957
XM_017017273.1
XP_016872762.1
22
5712
XM_017017274.1
XP_016872763.1
24
5907
XM_017017275.1
XP_016872764.1
21
5662
XM_017017276.1
XP_016872765.1
23
6385
XM_017017277.1
XP_016872766.1
23
5705
XM_017017278.1
XP_016872767.1
21
5609
XM_017017279.1
XP_016872768.1
23
5627
XM_017017280.1
XP_016872769.1
23
6304
XM_017017281.1
XP_016872770.1
22
5804
XM_017017283.1
XP_016872772.1
22
5609
XM_017017284.1
XP_016872773.1
22
5281
XM_017017285.1
XP_016872774.1
22
6231
XM_017017286.1
XP_016872775.1
20
5145
XM_017017287.1
XP_016872776.1
13
4165
XM_017017288.1
XP_016872777.1
12
4004
XM_017017289.1
XP_016872778.1
12
5579
XM_017017290.1
XP_016872779.1
12
4224
XM_017017291.1
XP_016872780.1
11
3971
XM_017017292.1
XP_016872781.1
11
5483
GenBank mRNA
AK094585.1
-
1
173
AK125151.1
-
1
1181
AK125152.1
-
1
704
BC104969.1
-
1
272
BC110844.1
-
1
527
BX537627.1
-
1
62
BX648386.1
-
1
1578
CR933674.1
-
11
4121
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
sensory perception of pain
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
GMP metabolic process
GDP metabolic process
receptor localization to synapse
guanylate kinase activity
protein binding
kinase binding
ionotropic glutamate receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0