Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164269.1 | NP_001157741.1 | 10 | 2125 | ||
NM_001164270.1 | NP_001157742.1 | 9 | 2155 | |||
NM_002875.4 | NP_002866.2 | 10 | 2277 | |||
NM_133487.3 | NP_597994.3 | 10 | 2280 | |||
XM_006720626.3 | XP_006720689.1 | 10 | 2328 | |||
XM_011521857.2 | XP_011520159.1 | 10 | 2027 | |||
XM_011521858.2 | XP_011520160.1 | 10 | 2074 | |||
XM_011521859.2 | XP_011520161.1 | 10 | 2211 | |||
XM_011521860.2 | XP_011520162.1 | 10 | 2122 | |||
XM_011521861.2 | XP_011520163.1 | 9 | 1905 | |||
GenBank mRNA | D14134.1 | - | 10 | 2209 |