Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023967.1 | NP_076457.1 | 6 | 840 | ||
XM_006235041.2 | XP_006235103.1 | 4 | 2397 | |||
XM_006235043.2 | XP_006235105.1 | 5 | 2313 | |||
XM_006235044.2 | XP_006235106.1 | 3 | 2262 | |||
XM_008762197.1 | XP_008760419.1 | 5 | 1837 | |||
GenBank mRNA | AJ294476.1 | - | 6 | 840 |