Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130040.1 | NP_001123512.1 | 12 | 3167 | ||
NM_001130041.1 | NP_001123513.1 | 13 | 2852 | |||
NM_001202859.1 | NP_001189788.1 | 13 | 2852 | |||
NM_003029.4 | NP_003020.2 | 13 | 2855 | |||
NM_183001.4 | NP_892113.4 | 12 | 3164 | |||
XM_011509892.2 | XP_011508194.1 | 12 | 4407 | |||
XM_011509893.2 | XP_011508195.1 | 12 | 4405 | |||
XM_011509894.2 | XP_011508196.1 | 11 | 4357 | |||
XM_011509897.1 | XP_011508199.1 | 12 | 2792 | |||
XM_017002081.1 | XP_016857570.1 | 11 | 4325 | |||
XM_017002082.1 | XP_016857571.1 | 11 | 4323 | |||
XM_017002083.1 | XP_016857572.1 | 12 | 2762 | |||
GenBank mRNA | AB208849.1 | - | 8 | 3892 | ||
AK096169.1 | - | 12 | 3150 | |||
BC014158.2 | - | 13 | 2736 | 4541216 | ||
BC033925.1 | - | 10 | 2566 | 4896385 | ||
BX647136.1 | - | 13 | 3080 | |||
BX647149.1 | - | 12 | 3167 | |||
U73377.1 | - | 12 | 3141 | |||
X68148.1 | - | 13 | 2698 |
Gene Symbol: LOC101928120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HY343691.1 | - | 1 | 44 |