Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013179.2 | NP_037311.1 | 2 | 482 | ||
GenBank mRNA | AF019565.1 | - | 2 | 487 | ||
AF041241.1 | - | 2 | 486 |