Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326542.1 | NP_001313471.1 | 3 | 825 | ||
NM_001326543.1 | NP_001313472.1 | 3 | 796 | |||
NM_003112.4 | NP_003103.2 | 3 | 830 | |||
XM_005249828.2 | XP_005249885.1 | 3 | 784 | |||
XM_005249829.3 | XP_005249886.1 | 3 | 764 | |||
XM_011515486.2 | XP_011513788.1 | 3 | 764 | |||
XM_011515487.2 | XP_011513789.1 | 3 | 764 | |||
XM_011515489.2 | XP_011513791.1 | 3 | 764 | |||
XM_017012557.1 | XP_016868046.1 | 3 | 764 | |||
GenBank mRNA | AK289728.1 | - | 3 | 830 | ||
AK296749.1 | - | 4 | 751 | |||
BC109300.1 | - | 3 | 736 | 40034749 | ||
BC109301.1 | - | 3 | 702 | 40034751 | ||
X68561.1 | - | 3 | 793 |