Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031136.1 | NP_112398.1 | 2 | 134 | ||
GenBank mRNA | BC058137.1 | - | 2 | 148 | 6889807 | |
BC084720.1 | - | 2 | 123 | |||
BC097308.1 | - | 2 | 133 | |||
FQ211846.1 | - | 2 | 141 | |||
FQ211950.1 | - | 2 | 144 | |||
FQ213262.1 | - | 2 | 141 | |||
FQ214330.1 | - | 2 | 141 | |||
FQ217154.1 | - | 2 | 141 | |||
FQ217532.1 | - | 2 | 141 | |||
FQ217617.1 | - | 2 | 141 | |||
FQ217622.1 | - | 2 | 141 | |||
FQ220985.1 | - | 2 | 141 | |||
FQ221085.1 | - | 2 | 141 | |||
FQ221124.1 | - | 2 | 141 | |||
FQ221299.1 | - | 2 | 141 | |||
FQ221364.1 | - | 2 | 141 | |||
FQ221505.1 | - | 2 | 141 | |||
FQ221785.1 | - | 2 | 141 | |||
FQ221964.1 | - | 2 | 141 | |||
FQ222011.1 | - | 2 | 141 | |||
FQ222095.1 | - | 2 | 141 | |||
FQ222205.1 | - | 2 | 141 | |||
FQ222251.1 | - | 2 | 141 | |||
FQ222334.1 | - | 2 | 141 | |||
FQ222463.1 | - | 2 | 141 | |||
FQ222519.1 | - | 2 | 141 | |||
FQ222755.1 | - | 2 | 141 | |||
FQ223266.1 | - | 2 | 138 | |||
FQ223325.1 | - | 2 | 141 | |||
FQ228380.1 | - | 2 | 141 | |||
FQ228401.1 | - | 2 | 141 | |||
FQ228412.1 | - | 2 | 141 | |||
FQ228477.1 | - | 2 | 141 | |||
FQ229728.1 | - | 2 | 142 | |||
K01334.1 | - | 1 | 102 | |||
M26759.1 | - | 1 | 108 | |||
M34043.1 | - | 2 | 134 |