siRNA Details
Gene Symbol: TCF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134851.2 | NP_001128323.2 | 9 | 1836 | ||
NM_003202.3 | NP_003193.2 | 10 | 1797 | |||
NM_201632.3 | NP_963963.1 | 9 | 1353 | |||
NM_201634.3 | NP_963965.1 | 10 | 1449 | |||
NM_213648.3 | NP_998813.1 | 8 | 1587 | |||
NR_033449.1 | 8 | 1588 | ||||
XM_006714680.3 | XP_006714743.1 | 11 | 1898 | |||
XM_006714682.2 | XP_006714745.1 | 11 | 1921 | |||
XM_006714684.2 | XP_006714747.1 | 10 | 1828 | |||
XM_011543612.2 | XP_011541914.1 | 8 | 1299 | |||
XM_017009789.1 | XP_016865278.1 | 10 | 1466 | |||
XM_017009790.1 | XP_016865279.1 | 9 | 2325 | |||
GenBank mRNA | AK057580.1 | - | 8 | 1589 | ||
AK093530.1 | - | 8 | 1458 | |||
AK093683.1 | - | 5 | 1687 | |||
AK131428.1 | - | 6 | 1879 | |||
AK310591.1 | - | 8 | 1299 | |||
AL832816.1 | - | 10 | 1516 | |||
AL834166.1 | - | 8 | 4106 | |||
BC048769.1 | - | 10 | 1797 | 6063270 | ||
BC072023.1 | - | 10 | 1732 | 6292932 | ||
X59870.1 | - | 9 | 1333 | |||
X59871.1 | - | 10 | 1429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
