Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173134.2 | NP_775157.2 | 33 | 5141 | ||
XM_006238273.2 | XP_006238335.1 | 34 | 5626 | |||
XM_006238274.2 | XP_006238336.1 | 34 | 5620 | |||
XM_006238276.2 | XP_006238338.1 | 33 | 5528 | |||
XM_006238277.2 | XP_006238339.1 | 33 | 5522 | |||
XM_006238278.2 | XP_006238340.1 | 33 | 5375 | |||
XM_006238279.2 | XP_006238341.1 | 33 | 5368 | |||
XM_006238280.2 | XP_006238342.1 | 32 | 5297 | |||
XM_008763760.1 | XP_008761982.1 | 34 | 5626 | |||
XM_008763761.1 | XP_008761983.1 | 33 | 5495 | |||
GenBank mRNA | AF177478.1 | - | 2 | 179 | ||
JX524852.1 | - | 34 | 5234 |