Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172317.2 | NP_758528.1 | 6 | 229 | ||
XM_006228818.2 | XP_006228880.1 | 6 | 479 | |||
XM_006228819.2 | XP_006228881.1 | 6 | 333 | |||
XM_006228820.2 | XP_006228882.1 | 6 | 469 | |||
GenBank mRNA | AW142066.1 | - | 5 | 234 | ||
DY471861.1 | - | 6 | 601 |