Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003420.3 | NP_003411.3 | 4 | 2620 | ||
XM_017007129.1 | XP_016862618.1 | 4 | 2594 | |||
XM_017007130.1 | XP_016862619.1 | 4 | 2615 | |||
XM_017007131.1 | XP_016862620.1 | 4 | 2616 | |||
GenBank mRNA | BC013597.1 | - | 4 | 2598 | 4186299 | |
BC015892.1 | - | 3 | 2452 |