Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175760.2 | NP_786936.1 | 215 | |||
GenBank mRNA | M33936.1 | - | 215 |
Gene Symbol: Cyp4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC098705.1 | - | 1, 2 | 214 | 7106510 |