Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173329.2 | NP_775451.2 | 3 | 437 | ||
XM_006253367.1 | XP_006253429.1 | 3 | 481 | |||
GenBank mRNA | U16685.1 | - | 3 | 416 | ||
U16686.1 | - | 3 | 437 |