Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175583.1 | NP_783173.1 | 1 | 855 |
Set Membership: |
G-protein coupled receptor |