Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012590.2 | NP_036722.1 | 1 | 192 | ||
GenBank mRNA | BC083564.1 | - | 1 | 192 | 7112717 |