Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012519.2 | NP_036651.1 | 4 | 479 | ||
XM_006233284.2 | XP_006233346.1 | 4 | 698 | |||
XM_006233285.2 | XP_006233347.1 | 4 | 698 | |||
XM_008761452.1 | XP_008759674.1 | 4 | 698 | |||
GenBank mRNA | BC107562.1 | - | 341 | 7939424 | ||
J05072.1 | - | 479 |