Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003608.3 | NP_003599.2 | 2 | 701 | ||
GenBank mRNA | AK313797.1 | - | 2 | 676 | ||
BC035633.1 | - | 2 | 673 | 5218256 | ||
BC071715.1 | - | 2 | 635 | 5180030 | ||
EU883574.1 | - | 1 | 143 |