Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012522.2 | NP_036654.2 | 8, 9 | 963 | ||
GenBank mRNA | D01098.1 | - | 8, 9 | 964 | ||
FQ211441.1 | - | 8, 9 | 1022 | |||
M88344.1 | - | 8, 9 | 1112 | |||
M88345.1 | - | 5, 6 | 1376 | |||
M88346.1 | - | 7, 8 | 858 | |||
M88347.1 | - | 7, 8 | 858 |