Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144070.1 | NP_001137542.1 | 4, 5 | 568 | ||
NM_003786.3 | NP_003777.2 | 4, 5 | 568 | |||
XM_005257763.3 | XP_005257820.1 | 4, 5 | 582 | |||
XM_011525422.2 | XP_011523724.1 | 4, 5 | 582 | |||
XM_011525423.1 | XP_011523725.1 | 4, 5 | 582 | |||
XM_011525425.2 | XP_011523727.1 | 3, 4 | 345 | |||
XM_017025266.1 | XP_016880755.1 | 4, 5 | 582 | |||
GenBank mRNA | AB010887.1 | - | 4, 5 | 502 | ||
AB208954.1 | - | 4, 5 | 508 | |||
AF009670.1 | - | 4, 5 | 558 | |||
AF083552.1 | - | 4, 5 | 505 | |||
AF085690.1 | - | 4, 5 | 524 | |||
AF085691.1 | - | 4, 5 | 524 | |||
AF085692.1 | - | 4, 5 | 524 | |||
AF104943.1 | - | 4, 5 | 492 | |||
BC046126.1 | - | 4, 5 | 568 | |||
BC050370.1 | - | 4, 5 | 493 | |||
BC058907.1 | - | 4, 5 | 527 | |||
BC104952.1 | - | 4, 5 | 520 | |||
BC137347.1 | - | 4, 5 | 519 | 9021345 | ||
BC137348.1 | - | 4, 5 | 519 | 9021346 | ||
Y17151.2 | - | 4, 5 | 530 |