Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005246.1 | NP_001005246.1 | 3 | 199 | ||
XM_008773252.1 | XP_008771474.1 | 3 | 266 | |||
XM_008773253.1 | XP_008771475.1 | 3 | 266 | |||
GenBank mRNA | X65468.1 | - | 3 | 214 | ||
X69767.1 | - | 3 | 214 |