Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012641.1 | NP_036773.1 | 3 | 199 | ||
GenBank mRNA | M18962.1 | - | 2 | 175 | ||
M62930.1 | - | 3 | 199 |