Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012730.1 | NP_036862.1 | 3, 4 | 639 | ||
GenBank mRNA | AB008423.1 | - | 3, 4 | 515 | ||
BC078897.1 | - | 3, 4 | 574 | 7109496 | ||
FQ218496.1 | - | 3, 4 | 580 | |||
M16655.1 | - | 3, 4 | 565 | |||
M22330.1 | - | 3, 4 | 639 |