Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012822.1 | NP_036954.1 | 2 | 326 | ||
GenBank mRNA | J03960.1 | - | 2 | 326 |