Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012904.2 | NP_037036.1 | 3, 4 | 243 | ||
XM_006231159.1 | XP_006231221.1 | 4, 5 | 411 | |||
XM_008760295.1 | XP_008758517.1 | 3, 4 | 330 | |||
GenBank mRNA | BC061710.1 | - | 3, 4 | 241 | 5598112 | |
FQ213489.1 | - | 3, 4 | 258 | |||
FQ215033.1 | - | 3, 4 | 258 | |||
FQ216134.1 | - | 3, 4 | 258 | |||
FQ228716.1 | - | 3, 4 | 259 | |||
FQ229071.1 | - | 3, 4 | 259 | |||
FQ229198.1 | - | 2, 3 | 179 | |||
M19967.1 | - | 2, 3 | 210 | |||
Y00446.1 | - | 3, 4 | 244 |