Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012929.1 | NP_037061.1 | 3 | 261 | ||
XM_006242308.2 | XP_006242370.1 | 4 | 566 | |||
GenBank mRNA | L48440.1 | - | 3 | 261 |