Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003658.4 | NP_003649.2 | 3 | 612 | ||
XM_011543043.1 | XP_011541345.1 | 3 | 439 | |||
XM_011543044.2 | XP_011541346.1 | 3 | 457 | |||
GenBank mRNA | AF031924.1 | - | 3 | 656 | ||
AJ243512.1 | - | 3 | 578 | |||
BC069378.1 | - | 3 | 612 | 7262194 | ||
BC111432.1 | - | 3 | 581 | 40069348 | ||
BC111572.1 | - | 3 | 581 | 40069358 |