Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013022.2 | NP_037154.2 | 3 | 357 | ||
XM_006239909.2 | XP_006239971.1 | 3 | 387 | |||
XM_008764574.1 | XP_008762796.1 | 3 | 387 | |||
XM_008764575.1 | XP_008762797.1 | 3 | 387 | |||
GenBank mRNA | U38481.1 | - | 3 | 355 |