Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167840.1 | NP_001161312.1 | 9, 10 | 1229 | ||
NM_012968.1 | NP_037100.1 | 8, 9 | 1148 | |||
XM_006248504.2 | XP_006248566.1 | 8, 9 | 1217 | |||
GenBank mRNA | AB079119.1 | - | 625 | |||
GU123169.1 | - | 9, 10 | 1229 | |||
U48592.1 | - | 8, 9 | 1148 |