Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081955.2 | NP_001075424.1 | 14 | 1210 | ||
NM_001165933.1 | NP_001159405.1 | 14 | 1210 | |||
NM_003835.3 | NP_003826.2 | 14 | 1219 | |||
XM_011525426.2 | XP_011523728.1 | 7 | 1390 | |||
GenBank mRNA | AF071476.1 | - | 14 | 1083 | ||
AF073710.1 | - | 5 | 944 | |||
AF493932.1 | - | 4 | 329 | |||
AK289876.1 | - | 14 | 1083 | |||
AK290535.1 | - | 14 | 1079 | |||
AK301719.1 | - | 14 | 1089 | |||
AK309061.1 | - | 14 | 1100 | |||
AY585190.1 | - | 4 | 329 | |||
AY585191.1 | - | 14 | 1007 | |||
BC022504.1 | - | 14 | 1077 | 4801328 | ||
BC096843.1 | - | 4 | 332 | 6519202 |