Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013103.1 | NP_037235.1 | 2 | 397 | ||
GenBank mRNA | BC081826.1 | - | 2 | 512 | 7109422 | |
EF121973.1 | - | 2 | 367 | |||
EF121974.1 | - | 2 | 367 | |||
EF122006.1 | - | 2 | 367 | |||
X56546.1 | - | 2 | 397 |