Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CFLAR SIRNA Assays ›
Gene Symbol:
CFLAR
Gene Name:
CASP8 and FADD like apoptosis regulator
Gene Aliases:
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
Chromosome Location:
Chr.2: 201116015 - 201172688 on Build GRCh38
UniGene ID:
Hs.390736
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4916
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CFLAR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127183.2
NP_001120655.1
2
689
NM_001127184.2
NP_001120656.1
2
661
NM_001202516.1
NP_001189445.1
2
661
NM_001308042.1
NP_001294971.1
1
333
NM_001308043.1
NP_001294972.1
2
711
NM_003879.5
NP_003870.4
2
661
XM_017005189.1
XP_016860678.1
2
652
XM_017005190.1
XP_016860679.1
2
525
XM_017005191.1
XP_016860680.1
2
604
XM_017005192.1
XP_016860681.1
2
652
XM_017005193.1
XP_016860682.1
2
2669
XM_017005194.1
XP_016860683.1
2
525
XM_017005195.1
XP_016860684.1
2
642
XM_017005197.1
XP_016860686.1
2
2669
XM_017005198.1
XP_016860687.1
2
525
GenBank mRNA
AB209600.1
-
2
408
AF005774.1
-
2
631
AF005775.1
-
1
196
AF009616.1
-
2
608
AF009617.1
-
1
196
AF009618.1
-
1
196
AF009619.1
-
1
196
AF010127.1
-
2
606
AF015450.1
-
2
700
AF015451.1
-
1
196
AF015452.1
-
1
196
AF041458.1
-
1
211
AF041460.1
-
1
196
AF041461.1
-
1
151
AK289913.1
-
2
485
AK308767.1
-
2
571
AK315208.1
-
2
432
BC001602.1
-
2
455
3542993
BT006751.1
-
1
196
BX457155.2
-
2
678
U85059.1
-
1
196
U97074.1
-
2
578
U97075.1
-
1
489
Y14039.1
-
2
677
Y14040.1
-
2
677
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
apoptotic process
skeletal muscle tissue development
skeletal muscle atrophy
regulation of skeletal muscle satellite cell proliferation
skeletal myofibril assembly
viral process
negative regulation of apoptotic process
positive regulation of catalytic activity
positive regulation of I-kappaB kinase/NF-kappaB signaling
skeletal muscle tissue regeneration
positive regulation of NF-kappaB transcription factor activity
regulation of necroptotic process
execution phase of apoptosis
negative regulation of myoblast fusion
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway
protease binding
death receptor binding
protein binding
enzyme activator activity
protein complex binding
cysteine-type endopeptidase activity involved in execution phase of apoptosis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0