Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013128.1 | NP_037260.1 | 9 | 1470 | ||
GenBank mRNA | BC085762.1 | - | 9 | 1469 | 7193151 | |
J04625.1 | - | 9 | 1425 | |||
M31602.1 | - | 9 | 1441 | |||
X51406.1 | - | 9 | 1470 |