Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003972.2 | NP_003963.1 | 3 | 293 | ||
XM_011540326.1 | XP_011538628.1 | 3 | 503 | |||
XM_011540329.2 | XP_011538631.1 | 3 | 503 | |||
XM_017016877.1 | XP_016872366.1 | 3 | 502 | |||
XM_017016878.1 | XP_016872367.1 | 2 | 181 | |||
XM_017016879.1 | XP_016872368.1 | 2 | 181 | |||
XM_017016880.1 | XP_016872369.1 | 2 | 181 | |||
XM_017016881.1 | XP_016872370.1 | 2 | 181 | |||
GenBank mRNA | AF038362.1 | - | 3 | 312 | ||
AJ001017.2 | - | 3 | 207 | |||
AK308399.1 | - | 3 | 502 | |||
BC112201.1 | - | 3 | 277 | 8327669 |