Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161441.1 | NP_001154913.1 | 9 | 1384 | ||
NM_001161442.1 | NP_001154914.1 | 9 | 1323 | |||
NM_001161443.1 | NP_001154915.1 | 8 | 1233 | |||
NM_003975.3 | NP_003966.2 | 9 | 1354 | |||
XM_006711615.2 | XP_006711678.1 | 9 | 1372 | |||
XM_017002762.1 | XP_016858251.1 | 9 | 1401 | |||
XM_017002763.1 | XP_016858252.1 | 9 | 1392 | |||
XM_017002764.1 | XP_016858253.1 | 9 | 1339 | |||
XM_017002765.1 | XP_016858254.1 | 9 | 1352 | |||
XM_017002766.1 | XP_016858255.1 | 9 | 1342 | |||
XM_017002767.1 | XP_016858256.1 | 8 | 1119 | |||
GenBank mRNA | AF051325.1 | - | 9 | 1276 | ||
AF097744.1 | - | 9 | 1253 | |||
AJ000553.1 | - | 9 | 1300 | |||
AY763098.1 | - | 8 | 1233 | |||
AY763099.1 | - | 9 | 1330 | |||
AY763100.1 | - | 9 | 1260 | |||
BC012107.1 | - | 9 | 1316 | 4636736 |