Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AURKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256834.2 | NP_001243763.1 | 4 | 313 | ||
NM_001284526.1 | NP_001271455.1 | 5 | 389 | |||
NM_001313950.1 | NP_001300879.1 | 5 | 362 | |||
NM_001313951.1 | NP_001300880.1 | 5 | 391 | |||
NM_001313952.1 | NP_001300881.1 | 4 | 377 | |||
NM_001313954.1 | NP_001300883.1 | 5 | 386 | |||
NM_004217.3 | NP_004208.2 | 5 | 386 | |||
NR_132731.1 | 4 | 268 | ||||
XM_011524072.2 | XP_011522374.1 | 4 | 502 | |||
XM_017025307.1 | XP_016880796.1 | 4 | 429 | |||
XM_017025309.1 | XP_016880798.1 | 4 | 257 | |||
GenBank mRNA | AB011446.1 | - | 5 | 330 | ||
AB011450.1 | - | 5 | 321 | |||
AB519677.1 | - | 4 | 264 | |||
AB519679.1 | - | 4 | 267 | |||
AF004022.1 | - | 5 | 322 | |||
AF008552.1 | - | 4 | 264 | |||
AF015254.1 | - | 5 | 322 | |||
AK297976.1 | - | 4 | 268 | |||
AK314889.1 | - | 5 | 335 | |||
AY677083.1 | - | 4 | 268 | |||
BC000442.2 | - | 5 | 325 | 2820719 | ||
BC009751.2 | - | 5 | 305 | 3677349 | ||
BC013300.2 | - | 5 | 345 | |||
BC080581.1 | - | 5 | 317 | 2819846 | ||
BE514591.1 | - | 4 | 311 | |||
BM905813.1 | - | 4 | 338 | |||
BP225027.1 | - | 5 | 372 | |||
BT019534.1 | - | 4 | 264 | |||
BU181480.1 | - | 4 | 245 | |||
CX781462.1 | - | 5 | 298 |