Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003592.2 | NP_003583.2 | 5, 6 | 892 | ||
XM_005250060.4 | XP_005250117.1 | 5, 6 | 928 | |||
XM_011516629.2 | XP_011514931.1 | 5, 6 | 884 | |||
XM_011516630.2 | XP_011514932.1 | 5, 6 | 785 | |||
XM_011516631.2 | XP_011514933.1 | 5, 6 | 913 | |||
XM_011516632.2 | XP_011514934.1 | 5, 6 | 811 | |||
XM_017012723.1 | XP_016868212.1 | 5, 6 | 812 | |||
GenBank mRNA | AF062536.1 | - | 4, 5 | 666 | ||
AK096163.1 | - | 6, 7 | 780 | |||
AK296909.1 | - | 4, 5 | 617 | |||
AK315023.1 | - | 5, 6 | 1068 | |||
BC125119.1 | - | 5, 6 | 805 | 40118923 | ||
BC125120.1 | - | 5, 6 | 805 | 40118927 | ||
BX537409.1 | - | 5, 6 | 822 | |||
U58087.1 | - | 5, 6 | 594 |