Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017105.1 | NP_058801.1 | 1, 2 | 922 | ||
GenBank mRNA | D63860.1 | - | 1, 2 | 922 |