Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003620.3 | NP_003611.1 | 6 | 2077 | ||
GenBank mRNA | BC016480.1 | - | 6 | 1993 | 3923593 | |
BC033893.2 | - | 6 | 1951 | 5265521 | ||
BC042418.1 | - | 7 | 2102 | 5167004 | ||
BC060877.1 | - | 6 | 2077 | 5260081 | ||
U78305.1 | - | 6 | 2048 |