Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316361.2 | NP_001303290.1 | 5 | 1087 | ||
NM_001323347.1 | NP_001310276.1 | 5 | 800 | |||
NM_003657.3 | NP_003648.2 | 5 | 1087 | |||
XM_005260587.2 | XP_005260644.1 | 5 | 867 | |||
XM_005260589.2 | XP_005260646.1 | 5 | 867 | |||
XM_005260590.2 | XP_005260647.1 | 5 | 867 | |||
XM_005260591.2 | XP_005260648.1 | 5 | 867 | |||
XM_005260593.2 | XP_005260650.1 | 5 | 867 | |||
XM_005260594.2 | XP_005260651.1 | 5 | 867 | |||
XM_005260595.2 | XP_005260652.1 | 5 | 867 | |||
XM_011529090.2 | XP_011527392.1 | 5 | 795 | |||
XM_011529091.2 | XP_011527393.1 | 5 | 867 | |||
XM_017028110.1 | XP_016883599.1 | 5 | 867 | |||
XM_017028111.1 | XP_016883600.1 | 5 | 867 | |||
XM_017028112.1 | XP_016883601.1 | 5 | 867 | |||
GenBank mRNA | AF041260.1 | - | 5 | 866 | ||
AK225656.1 | - | 5 | 856 | |||
AK294150.1 | - | 1 | 173 | |||
AK295318.1 | - | 2 | 266 | |||
AK308384.1 | - | 5 | 855 | |||
AK311215.1 | - | 2 | 269 | |||
AK315220.1 | - | 5 | 1087 | |||
BC024044.1 | - | 5 | 868 | 4645428 | ||
BC126346.1 | - | 5 | 798 | 8992062 |