Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017115.2 | NP_058811.2 | 1 | 320 | ||
GenBank mRNA | AF054894.1 | - | 1 | 286 | ||
M24393.1 | - | 1 | 320 |